siRNA Details
| Gene Symbol: Xkr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173393.2 | NP_775569.2 | 3 | 1903 | ||
| GenBank mRNA | AY534252.1 | - | 3 | 1609 | ||
| BC024502.1 | - | 3 | 1459 | 4983341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr6| Gene Symbol: Xkr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173393.2 | NP_775569.2 | 3 | 1903 | ||
| GenBank mRNA | AY534252.1 | - | 3 | 1609 | ||
| BC024502.1 | - | 3 | 1459 | 4983341 | ||