siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146055.3 | NP_666167.3 | 15 | 2173 | ||
| GenBank mRNA | AK028194.1 | - | 15 | 2095 | ||
| AK031472.1 | - | 11 | 1713 | |||
| BC010499.1 | - | 1 | 214 | |||
| BC021475.1 | - | 15 | 2203 | 5343456 | ||
| BC025471.1 | - | 2 | 395 | |||
| BC038346.1 | - | 15 | 2125 | 3985250 | ||

Ccar2
Genomic Map