siRNA Details
| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027906.1 | NP_082182.1 | 14, 15 | 1797 | ||
| NM_173758.3 | NP_776119.2 | 14, 15 | 1797 | |||
| GenBank mRNA | AK033642.1 | - | 14, 15 | 1745 | ||
| AK154108.1 | - | 14, 15 | 1781 | |||
| BC019143.1 | - | 14, 15 | 1729 | 5042800 | ||

Vwa8
Genomic Map