siRNA Details
| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 4, 5 | 2526 | ||
| GenBank mRNA | AK085667.1 | - | 4, 5 | 2442 | ||
| BC117979.1 | - | 3, 4 | 2250 | 40099589 | ||
| BC117980.1 | - | 3, 4 | 2250 | 40099596 | ||
| CN692948.1 | - | 1, 2 | 81 | |||
| D90343.1 | - | 4, 5 | 2327 | |||
| X56304.1 | - | 3, 4 | 2260 | |||

Tnc
Genomic Map