siRNA Details
| Gene Symbol: Tnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011607.3 | NP_035737.2 | 24, 25 | 6178 | ||
| GenBank mRNA | BC117979.1 | - | 18, 19 | 4537 | 40099589 | |
| BC117980.1 | - | 18, 19 | 4537 | 40099596 | ||
| CD352874.1 | - | 3, 4 | 360 | |||
| D90343.1 | - | 24, 25 | 5979 | |||
| X56304.1 | - | 23, 24 | 5912 | |||

Tnc
Genomic Map