siRNA Details
| Gene Symbol: Tnfaip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166402.1 | NP_001159874.1 | 8 | 2266 | ||
| NM_009397.3 | NP_033423.3 | 9 | 2351 | |||
| GenBank mRNA | AK088297.1 | - | 9 | 2308 | ||
| AK151921.1 | - | 9 | 2312 | |||
| AK153238.1 | - | 9 | 2306 | |||
| AK172529.1 | - | 8 | 2268 | |||
| BC050259.1 | - | 7 | 4147 | |||
| BC054806.1 | - | 9 | 2351 | 6838303 | ||
| BC060221.1 | - | 9 | 2351 | 6843338 | ||
| CT010397.1 | - | 8 | 2078 | |||
| U19463.1 | - | 8 | 2078 | |||

Tnfaip3
Genomic Map