siRNA Details
| Gene Symbol: Tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020275.4 | NP_064671.2 | 8 | 1193 | ||
| GenBank mRNA | AB031081.1 | - | 8 | 1133 | ||
| AF176833.1 | - | 8 | 1005 | |||
| AK016138.1 | - | 4 | 760 | |||
| AK050753.1 | - | 8 | 1194 | |||
| AK080069.1 | - | 8 | 1184 | |||
| AK154993.1 | - | 8 | 1173 | |||
| BC065141.1 | - | 8 | 1136 | 30608306 | ||

Tnfrsf10b
Genomic Map