siRNA Details
| Gene Symbol: Tnks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175091.3 | NP_780300.2 | 10 | 1650 | ||
| GenBank mRNA | AK048860.1 | - | 6 | 1015 | ||
| AK080913.1 | - | 10 | 1653 | |||
| BC046509.1 | - | 131 | ||||
| BC057370.1 | - | 10 | 1631 | 6835784 | ||

Tnks
Genomic Map