siRNA Details
| Gene Symbol: Tnnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163664.1 | NP_001157136.1 | 12 | 389 | ||
| NM_001163665.1 | NP_001157137.1 | 12 | 389 | |||
| NM_001163666.1 | NP_001157138.1 | 10 | 362 | |||
| NM_001163667.1 | NP_001157139.1 | 11 | 377 | |||
| NM_001163668.1 | NP_001157140.1 | 10 | 338 | |||
| NM_001163669.1 | NP_001157141.1 | 11 | 350 | |||
| NM_001163670.1 | NP_001157142.1 | 8 | 305 | |||
| NM_011620.3 | NP_035750.2 | 9 | 317 | |||
| GenBank mRNA | AA028290.1 | - | 11 | 330 | ||
| AA771596.1 | - | 10 | 315 | |||
| AI173874.1 | - | 10 | 354 | |||
| AI574395.1 | - | 12 | 383 | |||
| AI746973.1 | - | 10 | 357 | |||
| BC003747.1 | - | 12 | 385 | 3594256 | ||
| BY766323.1 | - | 11 | 367 | |||
| CB196352.1 | - | 2 | 141 | |||
| CK791812.1 | - | 1 | 27 | |||
| DV049834.1 | - | 9 | 364 | |||
| L48988.1 | - | 6 | 208 | |||
| L48989.1 | - | 6 | 208 | |||
| L48990.1 | - | 9 | 253 | |||
| L48991.1 | - | 6 | 235 | |||
| L48992.1 | - | 8 | 241 | |||
| L48993.1 | - | 8 | 241 | |||
| L49018.1 | - | 6 | 235 | |||
| L49466.1 | - | 11 | 377 | |||
| L49467.1 | - | 12 | 389 | |||
| L49468.1 | - | 10 | 362 | |||
| L49469.1 | - | 10 | 362 | |||
| L49470.1 | - | 11 | 350 | |||
| L49471.1 | - | 10 | 359 | |||
| L49472.1 | - | 10 | 338 | |||
| U77779.1 | - | 9 | 310 | |||

Tnnt3
Genomic Map