siRNA Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009408.2 | NP_033434.2 | 14 | 1775 | ||
| GenBank mRNA | CF734074.1 | - | 2 | 177 | ||
| D10061.1 | - | 14 | 1442 | |||
| L20632.1 | - | 14 | 1439 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Top1| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009408.2 | NP_033434.2 | 14 | 1775 | ||
| GenBank mRNA | CF734074.1 | - | 2 | 177 | ||
| D10061.1 | - | 14 | 1442 | |||
| L20632.1 | - | 14 | 1439 | |||