siRNA Details
| Gene Symbol: Tpbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164792.1 | NP_001158264.1 | 2 | 1686 | ||
| NM_011627.4 | NP_035757.2 | 2 | 1795 | |||
| GenBank mRNA | AK049377.1 | - | 1 | 138 | ||
| AK051162.1 | - | 2 | 1766 | |||
| AK143519.1 | - | 2 | 1625 | |||
| AK165597.1 | - | 2 | 1798 | |||
| BC058198.1 | - | 2 | 1654 | 5353871 | ||

Tpbg
Genomic Map