siRNA Details
| Gene Symbol: Tpst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130476.2 | NP_001123948.1 | 2 | 961 | ||
| NM_013837.2 | NP_038865.1 | 2 | 778 | |||
| GenBank mRNA | AF038008.1 | - | 2 | 778 | ||
| AK150753.1 | - | 2 | 846 | |||
| BC012666.1 | - | 2 | 708 | 4035106 | ||
| CT010202.1 | - | 1 | 568 | |||

Tpst1
Genomic Map