siRNA Details
| Gene Symbol: Traf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009421.3 | NP_033447.2 | 9 | 1224 | ||
| GenBank mRNA | AK029135.1 | - | 9 | 1226 | ||
| AK145075.1 | - | 799 | ||||
| AK169515.1 | - | 9 | 1017 | |||
| AK172598.1 | - | 5 | 828 | |||
| BC096595.1 | - | 5 | 790 | 30748947 | ||
| L35302.1 | - | 8 | 970 | |||

Traf1
Genomic Map