siRNA Details
| Gene Symbol: Traf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011633.2 | NP_035763.2 | 1364 | |||
| GenBank mRNA | AK144694.1 | - | 1315 | |||
| BC012702.1 | - | 1318 | 4218625 | |||
| D78141.1 | - | 1341 | ||||
| D83528.1 | - | 1206 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Traf5| Gene Symbol: Traf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011633.2 | NP_035763.2 | 1364 | |||
| GenBank mRNA | AK144694.1 | - | 1315 | |||
| BC012702.1 | - | 1318 | 4218625 | |||
| D78141.1 | - | 1341 | ||||
| D83528.1 | - | 1206 | ||||