siRNA Details
| Gene Symbol: Trhr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013696.2 | NP_038724.1 | 2 | 993 | ||
| GenBank mRNA | AK134591.1 | - | 2 | 994 | ||
| BC109005.1 | - | 2 | 878 | 40048176 | ||
| BC109006.1 | - | 2 | 878 | 40048178 | ||
| BC128269.1 | - | 2 | 878 | 40048433 | ||
| M59811.1 | - | 2 | 902 | |||
| M94384.1 | - | 2 | 902 | |||
More Information
Set Membership: |
|

Trhr
Genomic Map