siRNA Details
| Gene Symbol: Trpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011643.2 | NP_035773.1 | 9 | 2046 | ||
| GenBank mRNA | AF191551.1 | - | 8 | 1492 | ||
| AK046014.1 | - | 1 | 2477 | |||
| BC137581.1 | - | 9 | 2032 | 8860600 | ||
| BC144726.1 | - | 8 | 1930 | 9053274 | ||
| U73625.1 | - | 9 | 2046 | |||
| U95167.1 | - | 8 | 1944 | |||
| U95168.1 | - | 8 | 1891 | |||
| U95169.1 | - | 7 | 1789 | |||

Trpc1
Genomic Map