siRNA Details
| Gene Symbol: Trpc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282086.1 | NP_001269015.1 | 7 | 2091 | ||
| NM_013838.2 | NP_038866.2 | 8 | 2325 | |||
| GenBank mRNA | AF057748.1 | - | 8 | 1894 | ||
| AK133864.1 | - | 7 | 1991 | |||
| AK137447.1 | - | 7 | 1553 | |||
| BC141131.1 | - | 8 | 2196 | 9055743 | ||
| U49069.1 | - | 8 | 2325 | |||

Trpc6
Genomic Map