siRNA Details
| Gene Symbol: Tsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009440.2 | NP_033466.1 | 6, 7 | 499 | ||
| GenBank mRNA | AK018925.1 | - | 2, 3 | 140 | ||
| BC048493.1 | - | 6, 7 | 499 | 6705048 | ||
| X99796.1 | - | 6, 7 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsx| Gene Symbol: Tsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009440.2 | NP_033466.1 | 6, 7 | 499 | ||
| GenBank mRNA | AK018925.1 | - | 2, 3 | 140 | ||
| BC048493.1 | - | 6, 7 | 499 | 6705048 | ||
| X99796.1 | - | 6, 7 | 484 | |||