siRNA Details
| Gene Symbol: Tsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009440.2 | NP_033466.1 | 1 | 121 | ||
| GenBank mRNA | BC048493.1 | - | 1 | 121 | 6705048 | |
| HQ433540.1 | - | 2 | 232 | |||
| HQ433541.1 | - | 2 | 336 | |||
| HQ433542.1 | - | 2 | 231 | |||
| HQ433544.1 | - | 1 | 421 | |||
| HQ433547.1 | - | 1 | 321 | |||
| X99796.1 | - | 1 | 106 | |||

Tsx
Genomic Map