siRNA Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009441.2 | NP_033467.2 | 24 | 2324 | ||
| GenBank mRNA | AB008516.1 | - | 23 | 2201 | ||
| AK084201.1 | - | 14 | 1359 | |||
| AK088273.1 | - | 23 | 2271 | |||
| AK165850.1 | - | 13 | 1232 | |||
| AK166120.1 | - | 14 | 1264 | |||
| AK166164.1 | - | 15 | 1413 | |||
| AK220495.1 | - | 24 | 2366 | |||
| BC019173.1 | - | 24 | 2293 | |||
| BC037654.1 | - | 23 | 2224 | |||
| BC057207.1 | - | 23 | 2228 | |||
| BC145489.1 | - | 19 | 1844 | |||

Ttc3
Genomic Map