siRNA Details
| Gene Symbol: Ttk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110265.1 | NP_001103735.1 | 11, 12 | 1311 | ||
| NM_001284272.1 | NP_001271201.1 | 11, 12 | 1311 | |||
| NM_009445.2 | NP_033471.2 | 11, 12 | 1311 | |||
| GenBank mRNA | AK041487.1 | - | 11, 12 | 1282 | ||
| AK049200.1 | - | 11, 12 | 1306 | |||
| AK164093.1 | - | 11, 12 | 1310 | |||
| AX775731.1 | - | 11, 12 | 1277 | |||
| BC058851.1 | - | 11, 12 | 1297 | 30023533 | ||
| M86377.1 | - | 12, 13 | 1337 | |||

Ttk
Genomic Map