siRNA Details
| Gene Symbol: Ttk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110265.1 | NP_001103735.1 | 10, 11 | 1182 | ||
| NM_001284272.1 | NP_001271201.1 | 10, 11 | 1182 | |||
| NM_009445.2 | NP_033471.2 | 10, 11 | 1182 | |||
| GenBank mRNA | AK041487.1 | - | 10, 11 | 1153 | ||
| AK049200.1 | - | 10, 11 | 1177 | |||
| AK164093.1 | - | 10, 11 | 1181 | |||
| AX775731.1 | - | 10, 11 | 1148 | |||
| BC058851.1 | - | 10, 11 | 1168 | 30023533 | ||
| DV648812.1 | - | 7, 8 | 668 | |||
| M86377.1 | - | 11, 12 | 1208 | |||

Ttk
Genomic Map