siRNA Details
| Gene Symbol: Tuft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293728.1 | NP_001280657.1 | 2, 3 | 240 | ||
| NM_011656.3 | NP_035786.1 | 3, 4 | 315 | |||
| GenBank mRNA | AF002860.1 | - | 3, 4 | 209 | ||
| AF047704.1 | - | 3, 4 | 249 | |||
| AK132488.1 | - | 3, 4 | 247 | |||
| AK171705.1 | - | 2, 3 | 205 | |||
| BC019213.1 | - | 3, 4 | 246 | 5064658 | ||

Tuft1
Genomic Map