siRNA Details
| Gene Symbol: Zrsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011663.3 | NP_035793.1 | 1 | 1957 | ||
| GenBank mRNA | AK030552.1 | - | 1 | 747 | ||
| BC132555.1 | - | 2 | 1015 | 40130832 | ||
| BC138594.1 | - | 1 | 1025 | 8861614 | ||
| D17407.1 | - | 2 | 1902 | |||
| S69507.1 | - | 1 | 1891 | |||
| Gene Symbol: Commd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB076722.1 | - | 2 | 818 | ||

Zrsr1
Genomic Map