siRNA Details
| Gene Symbol: Zrsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011663.3 | NP_035793.1 | 1 | 1357 | ||
| GenBank mRNA | AK030552.1 | - | 1 | 147 | ||
| AK165562.1 | - | 2 | 1275 | |||
| BC132555.1 | - | 2 | 415 | 40130832 | ||
| BC138594.1 | - | 1 | 425 | 8861614 | ||
| D17407.1 | - | 2 | 1302 | |||
| S69507.1 | - | 1 | 1291 | |||

Zrsr1
Genomic Map