siRNA Details
| Gene Symbol: U2af2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205231.1 | NP_001192160.1 | 11 | 1369 | ||
| NM_133671.3 | NP_598432.2 | 11 | 1357 | |||
| GenBank mRNA | AK078496.1 | - | 11 | 1323 | ||
| BC007487.1 | - | 7 | 1591 | 3593924 | ||
| BC043071.1 | - | 11 | 1357 | 6403830 | ||
| BC094451.1 | - | 9 | 1010 | |||
| BC106134.1 | - | 10 | 1073 | 6309073 | ||
| X64587.1 | - | 11 | 1275 | |||

U2af2
Genomic Map