siRNA Details
| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009454.2 | NP_033480.1 | 5 | 855 | ||
| GenBank mRNA | AF003346.1 | - | 4 | 471 | ||
| AK076011.1 | - | 5 | 662 | |||
| AK168072.1 | - | 5 | 680 | |||
| AK184486.1 | - | 4 | 358 | |||
| AK189528.1 | - | 4 | 403 | |||
| BC011477.1 | - | 5 | 855 | 4195671 | ||
| X92664.1 | - | 5 | 639 | |||

Ube2e3
Genomic Map