siRNA Details
Gene Symbol: Ube2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009455.3 | NP_033481.1 | 4 | 421 | ||
GenBank mRNA | AK090177.1 | - | 4 | 422 | ||
AK148266.1 | - | 4 | 352 | |||
BC003781.1 | - | 4 | 338 | 3484294 | ||
X92665.1 | - | 4 | 383 |
