siRNA Details
| Gene Symbol: Usp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011669.3 | NP_035799.1 | 9 | 1349 | ||
| GenBank mRNA | AF441835.1 | - | 8 | 1010 | ||
| AK006739.1 | - | 9 | 1399 | |||
| AK035629.1 | - | 9 | 1190 | |||
| AK167153.1 | - | 9 | 1189 | |||
| BC023858.1 | - | 3 | 182 | |||
| BC049274.1 | - | 7 | 1016 | 4500832 | ||
| BC055398.1 | - | 5 | 441 | |||
| BC068136.1 | - | 9 | 1349 | 30363916 | ||

Usp12
Genomic Map