siRNA Details
Gene Symbol: Ubr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009461.2 | NP_033487.2 | 33 | 3849 | ||
GenBank mRNA | AF061555.1 | - | 33 | 3850 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ubr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009461.2 | NP_033487.2 | 33 | 3849 | ||
GenBank mRNA | AF061555.1 | - | 33 | 3850 |