siRNA Details
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013700.1 | NP_038728.1 | 13 | 1688 | ||
| GenBank mRNA | AK154008.1 | - | 13 | 1677 | ||
| AK179736.1 | - | 1 | 109 | |||
| AK194987.1 | - | 1 | 109 | |||
| BC066993.1 | - | 13 | 1666 | 5689229 | ||

Usp5
Genomic Map