siRNA Details
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013700.1 | NP_038728.1 | 7 | 875 | ||
| GenBank mRNA | AK154008.1 | - | 7 | 864 | ||
| BC066993.1 | - | 7 | 853 | 5689229 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp5| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013700.1 | NP_038728.1 | 7 | 875 | ||
| GenBank mRNA | AK154008.1 | - | 7 | 864 | ||
| BC066993.1 | - | 7 | 853 | 5689229 | ||