siRNA Details
| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011673.3 | NP_035803.1 | 7, 8 | 916 | ||
| GenBank mRNA | BC050828.1 | - | 7, 8 | 916 | 6512920 | |
| D89866.1 | - | 7, 8 | 1007 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugcg| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011673.3 | NP_035803.1 | 7, 8 | 916 | ||
| GenBank mRNA | BC050828.1 | - | 7, 8 | 916 | 6512920 | |
| D89866.1 | - | 7, 8 | 1007 | |||