siRNA Details
| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011673.3 | NP_035803.1 | 6 | 807 | ||
| GenBank mRNA | AK138848.1 | - | 6 | 822 | ||
| BC050828.1 | - | 6 | 807 | 6512920 | ||
| D89866.1 | - | 6 | 898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugcg| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011673.3 | NP_035803.1 | 6 | 807 | ||
| GenBank mRNA | AK138848.1 | - | 6 | 822 | ||
| BC050828.1 | - | 6 | 807 | 6512920 | ||
| D89866.1 | - | 6 | 898 | |||