siRNA Details
| Gene Symbol: Ugt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011674.4 | NP_035804.2 | 6 | 1560 | ||
| GenBank mRNA | AK128994.1 | - | 6 | 1556 | ||
| AK137364.1 | - | 6 | 1561 | |||
| AK144005.1 | - | 6 | 1560 | |||
| BC016885.1 | - | 1555 | 4223057 | |||
| X92122.1 | - | 5 | 1885 | |||

Ugt8a
Genomic Map