siRNA Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009469.3 | NP_033495.2 | 24, 25 | 3054 | ||
| GenBank mRNA | AF053756.1 | - | 24, 25 | 2746 | ||
| AF072370.1 | - | 24, 25 | 2737 | |||
| AK122357.1 | - | 23, 24 | 2556 | |||
| AK148317.1 | - | 24, 25 | 3047 | |||
| AK154939.1 | - | 24, 25 | 3071 | |||
| AK156534.1 | - | 24, 25 | 3031 | |||
| BC030850.1 | - | 3, 4 | 526 | |||
| BC053062.1 | - | 24, 25 | 2714 | |||
| BC057121.1 | - | 24, 25 | 3054 | 6834534 | ||
| BC059835.1 | - | 24, 25 | 3023 | 6406755 | ||

Ulk1
Genomic Map