siRNA Details
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135314.1 | - | 8 | 1264 | ||
| AK148116.1 | - | 9 | 959 | |||
| BC068161.1 | - | 9 | 765 | 30358504 | ||
| U04439.1 | - | 8 | 686 | |||
| U16216.1 | - | 9 | 827 | |||
| U18867.2 | - | 9 | 831 | |||

Uros
Genomic Map