siRNA Details
| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009481.2 | NP_033507.2 | 10 | 1623 | ||
| GenBank mRNA | AK028443.1 | - | 10 | 1608 | ||
| AK049283.1 | - | 11 | 1736 | |||
| DQ086491.1 | - | 10 | 1584 | |||
| U67874.1 | - | 10 | 1584 | |||

Usp9x
Genomic Map