siRNA Details
Gene Symbol: Slc45a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053077.3 | NP_444307.1 | 4 | 1094 | ||
GenBank mRNA | AF360357.1 | - | 4 | 1002 | ||
AK029155.1 | - | 4 | 1095 | |||
AK089932.1 | - | 4 | 1065 | |||
AK148207.1 | - | 1092 | ||||
AY034377.1 | - | 4 | 1015 | |||
BC125376.1 | - | 4 | 1081 | 40129891 | ||
BC132429.1 | - | 4 | 1081 | 40130706 |
