siRNA Details
| Gene Symbol: Slc45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053077.3 | NP_444307.1 | 1, 2 | 479 | ||
| GenBank mRNA | AF360357.1 | - | 1, 2 | 387 | ||
| AK029155.1 | - | 1, 2 | 480 | |||
| AK089932.1 | - | 1, 2 | 450 | |||
| AK148207.1 | - | 476 | ||||
| AY034377.1 | - | 1, 2 | 400 | |||
| BC125376.1 | - | 1, 2 | 466 | 40129891 | ||
| BC132429.1 | - | 1, 2 | 466 | 40130706 | ||

Slc45a2
Genomic Map