siRNA Details
| Gene Symbol: March6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172606.2 | NP_766194.2 | 15 | 1479 | ||
| GenBank mRNA | AK028353.1 | - | 1 | 13 | ||
| AK089827.1 | - | 15 | 1481 | |||
| AK129169.1 | - | 3 | 224 | |||
| AK132461.1 | - | 15 | 1452 | |||
| BC037454.1 | - | 1 | 33 | |||
| BC048816.1 | - | 2 | 119 | |||
| CF540592.1 | - | 9 | 541 | |||

March6
Genomic Map