siRNA Details
| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009511.2 | NP_033537.1 | 9 | 973 | ||
| GenBank mRNA | AK038743.1 | - | 9 | 974 | ||
| BC138572.1 | - | 9 | 871 | 8861592 | ||
| D28132.1 | - | 9 | 905 | |||
More Information
Other Species Targeted
Set Membership: |
|

Vipr2
Genomic Map