siRNA Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161420.1 | NP_001154892.1 | 17 | 2961 | ||
| NM_013703.2 | NP_038731.2 | 18 | 3045 | |||
| GenBank mRNA | AK044683.1 | - | 9 | 1569 | ||
| AK076172.1 | - | 17 | 2966 | |||
| AK136838.1 | - | 16 | 4109 | |||
| BC013622.1 | - | 17 | 2397 | 3968213 | ||
| L33417.1 | - | 18 | 2469 | |||
| U06670.1 | - | 18 | 2631 | |||

Vldlr
Genomic Map