siRNA Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161420.1 | NP_001154892.1 | 8 | 1752 | ||
| NM_013703.2 | NP_038731.2 | 8 | 1752 | |||
| GenBank mRNA | AK076172.1 | - | 8 | 1757 | ||
| AK136838.1 | - | 8 | 1572 | |||
| BC013622.1 | - | 8 | 1188 | 3968213 | ||
| L33417.1 | - | 8 | 1176 | |||
| U06670.1 | - | 8 | 1338 | |||

Vldlr
Genomic Map