siRNA Details
| Gene Symbol: Mroh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175457.4 | NP_780666.3 | 38 | 4338 | ||
| GenBank mRNA | AK052153.1 | - | 37 | 4292 | ||
| AK090131.1 | - | 33 | 8906 | |||
| AK147897.1 | - | 38 | 4328 | |||
| AK170702.1 | - | 38 | 4331 | |||
| BC039227.1 | - | 19 | 3330 | |||

Mroh1
Genomic Map