siRNA Details
| Gene Symbol: Mroh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175457.4 | NP_780666.3 | 39 | 4452 | ||
| GenBank mRNA | AK052153.1 | - | 38 | 4406 | ||
| AK090131.1 | - | 34 | 9020 | |||
| AK147897.1 | - | 39 | 4442 | |||
| AK170702.1 | - | 39 | 4445 | |||
| BC039227.1 | - | 20 | 3444 | |||

Mroh1
Genomic Map