siRNA Details
| Gene Symbol: Mroh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175457.4 | NP_780666.3 | 36 | 4062 | ||
| GenBank mRNA | AK052153.1 | - | 35 | 4014 | ||
| AK090131.1 | - | 31 | 8630 | |||
| AK147897.1 | - | 36 | 4052 | |||
| AK170702.1 | - | 36 | 4055 | |||
| BC039227.1 | - | 17 | 3054 | |||

Mroh1
Genomic Map