siRNA Details
| Gene Symbol: Trpv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011706.2 | NP_035836.2 | 4 | 636 | ||
| GenBank mRNA | AB021665.1 | - | 4 | 718 | ||
| AK008689.1 | - | 2 | 161 | |||
| AK089004.1 | - | 5 | 651 | |||
| AK134052.1 | - | 3 | 949 | |||
| AK162841.1 | - | 3 | 510 | |||
| AK171222.1 | - | 3 | 559 | |||
| AK200065.1 | - | 2 | 224 | |||
| BC005415.1 | - | 3 | 583 | 3487527 | ||

Trpv2
Genomic Map