siRNA Details
| Gene Symbol: Trpv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011706.2 | NP_035836.2 | 11 | 1900 | ||
| GenBank mRNA | AB021665.1 | - | 11 | 1982 | ||
| AK008689.1 | - | 9 | 1425 | |||
| AK089004.1 | - | 12 | 1915 | |||
| BC005415.1 | - | 10 | 1847 | 3487527 | ||

Trpv2
Genomic Map