siRNA Details
| Gene Symbol: Wap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011709.5 | NP_035839.3 | 3, 4 | 416 | ||
| GenBank mRNA | AK164735.1 | - | 3, 4 | 417 | ||
| BC132280.1 | - | 3, 4 | 414 | 40130557 | ||
| BC132282.1 | - | 3, 4 | 414 | 40130559 | ||
| V00856.1 | - | 3, 4 | 404 | |||

Wap
Genomic Map